Assay Details
| Gene Symbol: Gdpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044238.1 | NP_001037703.1 | 2 | 251 | 70 | |
| GenBank mRNA | BC105620.1 | - | 2 | 251 | 7453591 | 70 |
More Information
Set Membership: |
|

Gdpd1
Genomic Map