Assay Details
| Gene Symbol: Myo1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012983.2 | NP_037115.2 | 15 - 16 | 2068 | 62 | |
| XM_008767949.1 | XP_008766171.1 | 15 - 16 | 3837 | 62 | ||
| GenBank mRNA | BC087027.1 | - | 15 - 16 | 2068 | 7106587 | 62 |
| X71997.1 | - | 15 - 16 | 2118 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myo1d
Genomic Map