Assay Details
| Gene Symbol: Eif4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199372.2 | NP_955404.1 | 5 - 6 | 535 | 67 | |
| XM_006246624.2 | XP_006246686.1 | 5 - 6 | 545 | 67 | ||
| GenBank mRNA | BC063812.1 | - | 5 - 6 | 539 | 6921521 | 67 |
| FQ212737.1 | - | 5 - 6 | 535 | 67 | ||
| FQ225967.1 | - | 5 - 6 | 537 | 67 | ||
| FQ228869.1 | - | 5 - 6 | 537 | 67 | ||
| FQ233130.1 | - | 5 - 6 | 537 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif4a1
Genomic Map