Assay Details
| Gene Symbol: Ddc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247816.2 | XP_006247878.1 | 2 - 3 | 2011 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006247816.2 transcripts.
Set Membership: |
|

Ddc8
Genomic Map