Assay Details
| Gene Symbol: Zc2hc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107661.1 | NP_001101131.1 | 4 - 5 | 425 | 105 | |
| XM_006232144.2 | XP_006232206.1 | 4 - 5 | 444 | 105 | ||
| GenBank mRNA | BC168976.1 | - | 4 - 5 | 387 | 9033370 | 105 |
More Information
Updated Mapping Information
Set Membership: |
|

Zc2hc1a
Genomic Map