Assay Details
Gene Symbol: Gtf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113570.1 | NP_001107042.1 | 2 - 3 | 455 | 103 | |
GenBank mRNA | AF391798.1 | - | 2 - 3 | 216 | 103 | |
CB616240.1 | - | 2 - 3 | 463 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
