Assay Details
Gene Symbol: Stag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173507.1 | NP_001166978.1 | 29 - 30 | 3736 | 63 | |
XM_006257503.2 | XP_006257565.1 | 29 - 30 | 4772 | 63 | ||
XM_008773521.1 | XP_008771743.1 | 29 - 30 | 3906 | 63 | ||
XM_008773522.1 | XP_008771744.1 | 29 - 30 | 3470 | 63 | ||
XM_008773523.1 | XP_008771745.1 | 29 - 30 | 4429 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
