Assay Details
| Gene Symbol: Etv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228814.2 | XP_006228876.1 | 4 - 5 | 1109 | 84 | |
| XM_006228815.2 | XP_006228877.1 | 4 - 5 | 1127 | 84 | ||
| XM_008759255.1 | XP_008757477.1 | 3 - 4 | 1330 | 84 | ||
More Information
Set Membership: |
|

Etv2
Genomic Map