Assay Details
Gene Symbol: Kcnc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759309.1 | XP_008757531.1 | 4 - 5 | 2622 | 123 | |
GenBank mRNA | AY179604.1 | - | 3 - 4 | 2395 | 123 | |
M84210.1 | - | 4 - 5 | 2425 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
