Assay Details
| Gene Symbol: Bckdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012782.1 | NP_036914.1 | 6 - 7 | 856 | 77 | |
| GenBank mRNA | BC089915.1 | - | 4 - 5 | 503 | 77 | |
| BC161819.1 | - | 6 - 7 | 861 | 9099948 | 77 | |
| J02827.1 | - | 6 - 7 | 840 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bckdha
Genomic Map