Assay Details
Gene Symbol: Tmem91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728716.4 | XP_002728762.3 | 1 - 2 | 1910 | 79 | |
XM_006228522.2 | XP_006228584.1 | 1 - 2 | 1910 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
