Assay Details
| Gene Symbol: Snrnp70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108483.1 | NP_001101953.1 | 5 - 6 | 536 | 64 | |
| XM_006229101.2 | XP_006229163.1 | 4 - 5 | 480 | 64 | ||
| XM_006229102.2 | XP_006229164.1 | 4 - 5 | 402 | 64 | ||
| GenBank mRNA | BC167051.1 | - | 4 - 5 | 318 | 9089731 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|

Snrnp70
Genomic Map