Assay Details
Gene Symbol: Syne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009283.1 | NP_001009283.1 | 3 - 4 | 535 | 66 | |
XM_006228866.2 | XP_006228928.1 | 4 - 5 | 983 | 66 | ||
XM_006228867.1 | XP_006228929.1 | 4 - 5 | 770 | 66 | ||
XM_006228868.2 | XP_006228930.1 | 3 - 4 | 839 | 66 | ||
GenBank mRNA | BC088237.1 | - | 3 - 4 | 535 | 7325652 | 66 |
Gene Symbol: LOC100911963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748821.2 | XP_003748869.1 | 3 - 4 | 626 | 66 | |
XM_003748822.2 | XP_003748870.1 | 4 - 5 | 770 | 66 | ||
XM_006228801.2 | XP_006228863.1 | 4 - 5 | 984 | 66 | ||
XM_006228802.2 | XP_006228864.1 | 3 - 4 | 840 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
