Assay Details
Gene Symbol: Syne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009283.1 | NP_001009283.1 | 4 - 5 | 780 | 116 | |
XM_006228866.2 | XP_006228928.1 | 5 - 6 | 1228 | 116 | ||
XM_006228867.1 | XP_006228929.1 | 5 - 6 | 1015 | 116 | ||
XM_006228868.2 | XP_006228930.1 | 4 - 5 | 1084 | 116 | ||
GenBank mRNA | BC088237.1 | - | 4 - 5 | 780 | 7325652 | 116 |
Gene Symbol: LOC100911963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748821.2 | XP_003748869.1 | 4 - 5 | 871 | 116 | |
XM_003748822.2 | XP_003748870.1 | 5 - 6 | 1015 | 116 | ||
XM_006228801.2 | XP_006228863.1 | 5 - 6 | 1229 | 116 | ||
XM_006228802.2 | XP_006228864.1 | 4 - 5 | 1085 | 116 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
