Assay Details
| Gene Symbol: Atp6v1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106660.1 | NP_001100130.1 | 1 - 2 | 233 | 64 | |
| GenBank mRNA | BC166422.1 | - | 1 - 2 | 153 | 9088893 | 64 |
| FQ221371.1 | - | 1 - 2 | 185 | 64 | ||
| FQ222881.1 | - | 1 - 2 | 147 | 64 | ||
| FQ227939.1 | - | 1 - 2 | 185 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atp6v1g1
Genomic Map