Assay Details
| Gene Symbol: Megf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107940.1 | NP_001101410.1 | 5 - 6 | 1532 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P101180 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P277340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50188 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P277340 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P101180 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P277340 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P101180 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P277340 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P101180 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P539148 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P277340 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90931 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93046 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10877636 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877631 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877635 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877633 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877634 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877632 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10877630 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500021933.rn.1 | TC0500002941.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500021934.rn.1 | TC0500002941.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10877630 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21139493 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367211 | - | - |

Megf9
Genomic Map