Assay Details
| Gene Symbol: Cryzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013044.1 | NP_001013062.1 | 6 - 7 | 362 | 71 | |
| XM_006248046.2 | XP_006248108.1 | 5 - 6 | 295 | 71 | ||
| GenBank mRNA | BC083853.1 | - | 6 - 7 | 362 | 7124012 | 71 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P17564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116880 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P17564 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P17564 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P17564 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P448693 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P347442 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17564 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29667 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85666 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86514 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102067 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10753192 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753191 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753193 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753197 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753187 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753196 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753190 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753186 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753195 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753189 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753194 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753188 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853090 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10853089 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10753185 | - | - |
| Rat Genome 230 2.0 Array | 1392506_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397878_at | - | - |
| Rat Genome 230 2.0 Array | 1392506_at | - | - |
| Rat Genome 230 2.0 Array | 1397878_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392506_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392506_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100007829.rn.1 | TC1100001315.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100007830.rn.1 | TC1100001315.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100007822.rn.1 | TC1100001315.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100007823.rn.1 | TC1100001315.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100007824.rn.1 | TC1100001315.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100007827.rn.1 | TC1100001315.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100007825.rn.1 | TC1100001315.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100007828.rn.1 | TC1100001315.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100007832.rn.1 | TC1100001315.rn.1 | RTA TC |
| Rat Expression Set 230 | 1397878_PM_at | - | - |
| Rat Expression Set 230 | 1397878_at | - | - |
| Rat Expression Set 230 | 1392506_at | - | - |
| Rat Expression Set 230 | 1392506_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10753185 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21573657 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350259 | - | - |

Cryzl1
Genomic Map