Assay Details
Gene Symbol: Ddit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109986.1 | NP_001103456.1 | 2 - 3 | 144 | 87 | |
XM_006241444.2 | XP_006241506.1 | 3 - 4 | 1322 | 87 | ||
GenBank mRNA | AW916370.1 | - | 2 - 3 | 144 | 87 | |
BC100664.1 | - | 2 - 3 | 107 | 7932840 | 87 | |
U36994.1 | - | 1 - 2 | 66 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC100664.1, NM_001109986.1, U36994.1, XM_006241444.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
