Assay Details
| Gene Symbol: S100a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232603.2 | XP_006232665.1 | 1 - 2 | 673 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006232603.2 transcripts.
Set Membership: |
|

S100a1
Genomic Map