Assay Details
| Gene Symbol: Lmna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002016.2 | NP_001002016.2 | 1 - 2 | 557 | 68 | |
| GenBank mRNA | DN934590.1 | - | 1 - 2 | 297 | 68 | |
| X66870.1 | - | 1 - 2 | 516 | 68 | ||
| X76297.1 | - | 1 - 2 | 279 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lmna
Genomic Map