Assay Details
| Gene Symbol: Tmem154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108553.1 | NP_001102023.1 | 1 - 2 | 161 | 86 | |
| XM_006232538.2 | XP_006232600.1 | 1 - 2 | 336 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem154
Genomic Map