Assay Details
| Gene Symbol: Tsacc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108556.1 | NP_001102026.1 | 2 - 3 | 125 | 69 | |
| XM_006232690.2 | XP_006232752.1 | 2 - 3 | 176 | 69 | ||
| XM_008761180.1 | XP_008759402.1 | 2 - 3 | 140 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tsacc
Genomic Map