Assay Details
| Gene Symbol: S100a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761271.1 | XP_008759493.1 | 2 - 3 | 1709 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008761271.1 transcripts.
Set Membership: |
|

S100a14
Genomic Map