Assay Details
Gene Symbol: Dusp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106624.2 | NP_001100094.1 | 4 - 5 | 696 | 59 | |
XM_008763374.1 | XP_008761596.1 | 5 - 6 | 1272 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
protein dephosphorylation
biological_process
dephosphorylation
peptidyl-tyrosine dephosphorylation
negative regulation of MAPK cascade
molecular_function
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
inactivation of MAPK activity
protein dephosphorylation
biological_process
dephosphorylation
peptidyl-tyrosine dephosphorylation
negative regulation of MAPK cascade
molecular_function
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P024052 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P853102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116590 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P853102 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P024052 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P024052 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P853102 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P024052 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P853102 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P504036 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49397 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10866403 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866407 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866405 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866404 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866402 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866406 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10866401 | - | - |
Rat Genome 230 2.0 Array | 1372354_at | - | - |
Rat Genome 230 2.0 Array | 1372354_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372354_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372354_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400027962.rn.1 | TC0400003935.rn.1 | RTA TC |
Rat Expression Set 230 | 1372354_at | - | - |
Rat Expression Set 230 | 1372354_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10866401 | - | - |
Rat Genome U34 Set | rc_AI176298_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21155879 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373390 | - | - |
