Assay Details
Gene Symbol: Timp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109393.1 | NP_001102863.1 | 3 - 4 | 720 | 94 | |
GenBank mRNA | FQ213131.1 | - | 3 - 4 | 457 | 94 | |
FQ214060.1 | - | 3 - 4 | 428 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
