Assay Details
| Gene Symbol: Cd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012705.1 | NP_036837.1 | 6 - 7 | 1206 | 144 | |
| XM_006237331.2 | XP_006237393.1 | 8 - 9 | 1469 | 144 | ||
| XM_006237333.2 | XP_006237395.1 | 6 - 7 | 1229 | 144 | ||
| XM_008763280.1 | XP_008761502.1 | 6 - 7 | 1186 | 144 | ||
| GenBank mRNA | M15768.1 | - | 6 - 7 | 1206 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd4
Genomic Map