Assay Details
Gene Symbol: Mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC086394.1 | - | 2 - 3 | 144 | 7303359 | 135 |
FQ224403.1 | - | 2 - 3 | 169 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
