Assay Details
| Gene Symbol: Gprc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079890.1 | NP_001073359.1 | 1 - 2 | 102 | 82 | |
| GenBank mRNA | BC128767.1 | - | 1 - 2 | 102 | 7124284 | 82 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC128767.1, NM_001079890.1 transcripts.
Set Membership: |
|

Gprc5a
Genomic Map