Assay Details
| Gene Symbol: Fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201421.1 | NP_958824.1 | 1 - 2 | 319 | 76 | |
| XM_006237688.2 | XP_006237750.1 | 1 - 2 | 308 | 76 | ||
| XM_008763402.1 | XP_008761624.1 | 1 - 2 | 298 | 76 | ||
| GenBank mRNA | AY426739.1 | - | 1 - 2 | 319 | 76 | |
| AY426740.1 | - | 1 - 2 | 319 | 76 | ||
| BC087696.1 | - | 1 - 2 | 216 | 7312267 | 76 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AY426739.1,AY426740.1,BC087696.1,NM_201421.1,XM_006237688.2,XM_008763402.1 transcripts.
Set Membership: |
|

Fgfr1op2
Genomic Map