Assay Details
Gene Symbol: Csgalnact2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106616.1 | NP_001100086.1 | 4 - 5 | 1365 | 109 | |
XM_006237181.2 | XP_006237243.1 | 4 - 5 | 1138 | 109 | ||
XM_006237182.2 | XP_006237244.1 | 4 - 5 | 1379 | 109 | ||
GenBank mRNA | FQ221326.1 | - | 4 - 5 | 1277 | 109 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
