Assay Details
| Gene Symbol: Foxj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109352.1 | NP_001102822.1 | 2 - 3 | 1114 | 74 | |
| XM_006237311.2 | XP_006237373.1 | 3 - 4 | 927 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Foxj2
Genomic Map