Assay Details
| Gene Symbol: Bid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022684.1 | NP_073175.1 | 2 - 3 | 156 | 109 | |
| GenBank mRNA | AF259503.1 | - | 2 - 3 | 156 | 109 | |
| D89804.1 | - | 112 | 109 | |||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF259503.1,D89804.1,NM_022684.1 transcripts.
Set Membership: |
|

Bid
Genomic Map