Assay Details
Gene Symbol: Emp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012843.2 | NP_036975.1 | 315 | 178 | ||
XM_006237515.2 | XP_006237577.1 | 424 | 178 | |||
XM_008763369.1 | XP_008761591.1 | 424 | 178 | |||
GenBank mRNA | BC087031.1 | - | 315 | 7192247 | 178 | |
Z54212.1 | - | 284 | 178 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
