Assay Details
| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108309.1 | NP_001101779.1 | 16 - 17 | 2445 | 67 | |
| GenBank mRNA | FQ212255.1 | - | 16 - 17 | 2355 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P336813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111247 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P336813 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P336813 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P336813 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P336813 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P745623 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99487 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15336 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10739870 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739873 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739869 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739867 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739874 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739878 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739875 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739881 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739868 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739880 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739877 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739872 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739885 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739876 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739882 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739871 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739879 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10739866 | - | - |
| Rat Genome 230 2.0 Array | 1397334_at | - | - |
| Rat Genome 230 2.0 Array | 1397946_at | - | - |
| Rat Genome 230 2.0 Array | 1397946_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388334_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397334_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388334_at | - | - |
| Affymetrix HT Rat Focus Array | 1397334_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397334_at | - | - |
| Affymetrix HT Rat Focus Array | 1388334_at | - | - |
| Affymetrix HT Rat Focus Array | 1388334_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397946_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397946_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000018000.rn.1 | TC1000001993.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388334_at | - | - |
| Rat Expression Set 230 | 1388334_PM_at | - | - |
| Rat Expression Set 230 | 1397334_at | - | - |
| Rat Expression Set 230 | 1397334_PM_at | - | - |
| Rat Expression Set 230 | 1397946_at | - | - |
| Rat Expression Set 230 | 1397946_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739866 | - | - |
| Rat Genome U34 Set | rc_AI176849_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21798267 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373455 | - | - |

Sec14l1
Genomic Map