Assay Details
Gene Symbol: LOC687707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145538.1 | NP_001139010.1 | 2 - 3 | 296 | 58 | |
GenBank mRNA | BF565552.1 | - | 2 - 3 | 303 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
