Assay Details
Gene Symbol: Arf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246525.2 | XP_006246587.1 | 1 - 2 | 89 | 57 | |
GenBank mRNA | BC061552.1 | - | 1 - 2 | 58 | 6889332 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC061552.1,XM_006246525.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
