Assay Details
Gene Symbol: Arl5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109046.1 | NP_001102516.1 | 4 - 5 | 396 | 69 | |
XM_006247396.2 | XP_006247458.1 | 4 - 5 | 589 | 69 | ||
XM_006247397.2 | XP_006247459.1 | 4 - 5 | 583 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
