Assay Details
| Gene Symbol: Spag7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107016.1 | NP_001100486.1 | 1 - 2 | 217 | 80 | |
| GenBank mRNA | FQ233279.1 | - | 1 - 2 | 89 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag7
Genomic Map