Assay Details
| Gene Symbol: Aloxe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105793.1 | NP_001099263.1 | 5 - 6 | 704 | 80 | |
| XM_006246617.2 | XP_006246679.1 | 6 - 7 | 1336 | 80 | ||
| XM_006246619.2 | XP_006246681.1 | 5 - 6 | 776 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aloxe3
Genomic Map