Assay Details
| Gene Symbol: Ddx42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107059.1 | NP_001100529.1 | 9 - 10 | 1276 | 91 | |
| XM_006247596.2 | XP_006247658.1 | 9 - 10 | 1230 | 91 | ||
| XM_006247597.2 | XP_006247659.1 | 8 - 9 | 1041 | 91 | ||
| GenBank mRNA | FQ221897.1 | - | 8 - 9 | 841 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx42
Genomic Map