Assay Details
Gene Symbol: Cnot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013856.1 | NP_001013878.1 | 12 - 13 | 1736 | 142 | |
GenBank mRNA | BC079308.1 | - | 12 - 13 | 1736 | 7134677 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of translation
positive regulation of cell proliferation
positive regulation of cytoplasmic mRNA processing body assembly
gene silencing by RNA
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
nuclear-transcribed mRNA catabolic process, no-go decay
RNA phosphodiester bond hydrolysis, exonucleolytic
positive regulation of ligand-dependent nuclear receptor transcription coactivator activity
RNA binding
exoribonuclease activity
poly(A)-specific ribonuclease activity
metal ion binding
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of translation
positive regulation of cell proliferation
positive regulation of cytoplasmic mRNA processing body assembly
gene silencing by RNA
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
nuclear-transcribed mRNA catabolic process, no-go decay
RNA phosphodiester bond hydrolysis, exonucleolytic
positive regulation of ligand-dependent nuclear receptor transcription coactivator activity
RNA binding
exoribonuclease activity
poly(A)-specific ribonuclease activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P485747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26424 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P485747 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P485747 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P485747 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P485747 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37616 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100984 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62781 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56841 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10742385 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742376 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742372 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742378 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742380 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742377 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742382 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742374 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742384 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742383 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742373 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742381 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742371 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742375 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10742370 | - | - |
Rat Genome 230 2.0 Array | 1373576_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373576_at | - | - |
Rat Genome 230 2.0 Array | 1382509_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382509_at | - | - |
Rat Genome 230 2.0 Array | 1383450_at | - | - |
Rat Genome 230 2.0 Array | 1383450_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383450_at | - | - |
Affymetrix HT Rat Focus Array | 1382509_at | - | - |
Affymetrix HT Rat Focus Array | 1382509_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383450_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000023790.rn.1 | TC1000002697.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000023795.rn.1 | TC1000002697.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000023794.rn.1 | TC1000002697.rn.1 | RTA TC |
Rat Expression Set 230 | 1383450_PM_at | - | - |
Rat Expression Set 230 | 1382509_at | - | - |
Rat Expression Set 230 | 1383450_at | - | - |
Rat Expression Set 230 | 1382509_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742370 | - | - |
Rat Genome U34 Set | rc_AI070241_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357809 | - | - |
