Assay Details
| Gene Symbol: Neurl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107013.1 | NP_001100483.1 | 21 - 22 | 3531 | 58 | |
| XM_006246712.1 | XP_006246774.1 | 21 - 22 | 3531 | 58 | ||
| XM_008767829.1 | XP_008766051.1 | 19 - 20 | 2950 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Neurl4
Genomic Map