Assay Details
| Gene Symbol: Prss38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246538.2 | XP_006246600.1 | 3 - 4 | 1168 | 122 | |
| XM_220559.8 | XP_220559.5 | 4 - 5 | 994 | 122 | ||
More Information
Set Membership: |
|

Prss38
Genomic Map