Assay Details
| Gene Symbol: Olr1413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214821.1 | NP_999986.1 | 1 | 631 | 81 | |
| GenBank mRNA | X89695.1 | - | 1 | 254 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olr1413
Genomic Map