Assay Details
Gene Symbol: Kctd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC168958.1 | - | 695 | 107 |
Gene Symbol: Tmem95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134799.1 | NP_001128271.1 | 2 - 3 | 233 | 107 | |
XM_006246813.2 | XP_006246875.1 | 2 - 3 | 389 | 107 | ||
XM_008767876.1 | XP_008766098.1 | 2 - 3 | 389 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
