Assay Details
Gene Symbol: Kctd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC168958.1 | - | 788 | 94 |
Gene Symbol: Tmem95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246813.2 | XP_006246875.1 | 4 - 5 | 482 | 94 | |
XM_008767876.1 | XP_008766098.1 | 4 - 5 | 482 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
