Assay Details
Gene Symbol: C1qbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019259.2 | NP_062132.2 | 4 - 5 | 572 | 65 | |
GenBank mRNA | BC070510.1 | - | 4 - 5 | 572 | 7100505 | 65 |
FQ212622.1 | - | 4 - 5 | 597 | 65 | ||
FQ214537.1 | - | 4 - 5 | 622 | 65 | ||
FQ219796.1 | - | 4 - 5 | 595 | 65 | ||
FQ220621.1 | - | 4 - 5 | 615 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
