Assay Details
| Gene Symbol: Zfp207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039020.1 | NP_001034109.1 | 5 - 6 | 672 | 114 | |
| XM_006247001.1 | XP_006247063.1 | 5 - 6 | 733 | 114 | ||
| XM_006247002.1 | XP_006247064.1 | 5 - 6 | 733 | 114 | ||
| XM_008768060.1 | XP_008766282.1 | 5 - 6 | 733 | 114 | ||
| XM_008768061.1 | XP_008766283.1 | 5 - 6 | 733 | 114 | ||
| GenBank mRNA | BC100269.1 | - | 5 - 6 | 672 | 7387460 | 114 |
| FQ221862.1 | - | 4 - 5 | 418 | 114 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp207
Genomic Map