Assay Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019338.1 | NP_062211.1 | 1209 | 94 | ||
| GenBank mRNA | CB605941.1 | - | 1097 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019338.1 | NP_062211.1 | 1209 | 94 | ||
| GenBank mRNA | CB605941.1 | - | 1097 | 94 | ||
Set Membership: |
|