Assay Details
| Gene Symbol: Eif4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100158.2 | NP_001093628.1 | 9 - 10 | 1039 | 97 | |
| GenBank mRNA | BC105875.1 | - | 9 - 10 | 984 | 7130292 | 97 |
More Information
Updated Mapping Information
Set Membership: |
|

Eif4a3
Genomic Map